Merck
Regioselective copolymerization of acryl sucrose monomers.
The Journal of organic chemistry (2004-10-23)
Maria Teresa Barros, Krasimira T Petrova, Ana Maria Ramos
ABSTRACT
1',2,3,3',4,4',6-Hepta-O-benzyl-sucrose has been prepared and selectively 6'-O-esterified with small alpha,beta-unsaturated acid derivatives. New chiral copolymers containing sucrose have been synthesized by radical polymerization, and some of their physical properties have been determined.
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
tert-Butyl(chloro)diphenylsilane, 98%
Sigma-Aldrich
Tetrabutylammonium iodide, reagent grade, 98%
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%