Merck
  • Glatiramer acetate reverses cognitive deficits from cranial-irradiated rat by inducing hippocampal neurogenesis.
Glatiramer acetate reverses cognitive deficits from cranial-irradiated rat by inducing hippocampal neurogenesis.
Journal of neuroimmunology (2014-04-10)
Fen He, Jun-Tao Zou, Qiong-Fang Zhou, Dao-Li Niu, Wei-Hua Jia
ABSTRACT
Patients received cranial-irradiation can be affected with cognitive deficits and decreasing hippocampal neurogenesis. In this work, we characterized the cognitive ability and immune-induced neurogenesis of the pre- and post-treated cranial-irradiated rats with Glatiramer acetate (GA), known as a weak CNS auto-antigen. The GA-treated rats displayed better cognitive abilities in Morris water maze (MWM). The numbers of Iba-I-positive microglia, BrdU(+)/DCX(+) cells and BrdU(+)/NeuN(+) cells in hippocampus increased, which are accompanied with increased IFN-γ and decreased IL-6, IL-4. Furthermore, GA reverted the Th1/Th2 balance. GA treatment can reverse the cognitive deficits caused by cranial irradiation through a mechanism that likely involves immunomodulation.
Sigma-Aldrich
Cotton Rat IL4 / Interleukin 4 ELISA Kit
Sigma-Aldrich
Mouse IL-4 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-4 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human IL-4 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Rat IL-4 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IL-4 ELISA Kit, for serum, plasma and cell culture supernatant