Merck
  • Synthesis of enantiopure glycidol derivatives via a one-pot two-step enzymatic cascade.
Synthesis of enantiopure glycidol derivatives via a one-pot two-step enzymatic cascade.
Organic & biomolecular chemistry (2014-12-23)
Yu-Chang Liu, Yan Liu, Zhong-Liu Wu
ABSTRACT
Styrene monooxygenase (SMO) can catalyze the kinetic resolution of secondary allylic alcohols to provide enantiopure glycidol derivatives. To overcome the low theoretical yield of kinetic resolution, we designed a one-pot two-step enzymatic cascade using prochiral α,β-unsaturated ketones as the substrates. An S-specific ketoreductase ChKRED03 was screened for the efficient bioreduction of the substrates to provide (S)-allylic alcohols, which underwent SMO-catalyzed epoxidation to achieve glycidol derivatives with contiguous stereogenic centers. Excellent enantioselectivity (ee > 99%) and diastereoselectivity (de > 99%) were achieved for the majority of the substrates, and product yields reached up to >99%.
Product Description
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Nitrilotriacetic acid, Sigma Grade, ≥99%
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
3-Chloropropiophenone, 98%
Sigma-Aldrich
Nitrilotriacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Supelco
Nitrilotriacetic acid, Pharmaceutical Secondary Standard; Certified Reference Material