Merck
  • The tyrosine kinase Lyn limits the cytokine responsiveness of plasma cells to restrict their accumulation in mice.
The tyrosine kinase Lyn limits the cytokine responsiveness of plasma cells to restrict their accumulation in mice.
Science signaling (2014-08-15)
Simona Infantino, Sarah A Jones, Jennifer A Walker, Mhairi J Maxwell, Amanda Light, Kristy O'Donnell, Evelyn Tsantikos, Victor Peperzak, Toby Phesse, Matthias Ernst, Fabienne Mackay, Margaret L Hibbs, Kirsten A Fairfax, David M Tarlinton
ABSTRACT
Maintenance of an appropriate number of plasma cells, long-lived antibody-producing cells that are derived from B cells, is essential for maintaining immunological memory while limiting disease. Plasma cell survival relies on extrinsic factors, the limited availability of which determines the size of the plasma cell population. Mice deficient in the nonreceptor tyrosine kinase Lyn are prone to an autoimmune disease that is characterized by inflammation and an excess of plasma cells (plasmacytosis). We demonstrated that the plasmacytosis was intrinsic to B cells and independent of inflammation. We also showed that Lyn attenuated signaling by signal transducer and activator of transcription 3 (STAT3) and STAT5 in response to the cytokines interleukin-6 (IL-6) and IL-3, respectively, in two previously uncharacterized plasma cell signaling pathways. Thus, in the absence of Lyn, the survival of plasma cells was improved, which enabled the plasma cells to become established in excess numbers in niches in vivo. These data identify Lyn as a key regulator of survival signaling in plasma cells, limiting plasma cell accumulation and autoimmune disease susceptibility.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Anti-SLC7A5 (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAFC
BIS-TRIS
SAFC
BIS-TRIS
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%