Merck
  • In situ gelling polyvalerolactone-based thermosensitive hydrogel for sustained drug delivery.
In situ gelling polyvalerolactone-based thermosensitive hydrogel for sustained drug delivery.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-06-17)
Gyan P Mishra, Reid Kinser, Igor H Wierzbicki, Raid G Alany, Adam W G Alani
ABSTRACT
Biodegradable poly(ethyleneglycol)-poly(valerolactone)-poly(ethyleneglycol) [PEG-PVL-PEG] copolymers were synthesized through ring opening polymerization of δ-valerolactone (VL) followed by the coupling of monomethoxy poly(ethyleneglycol-poly(valerolactone) (mPEG-PVL) with hexamethylene diisocyanate (HDI). The copolymers were characterized by (1)H NMR, FT-IR, and GPC. Block copolymers of PEG and PVL with different VL/PEG molar ratios were successfully synthesized. One of the copolymers (Copolymer 2, PEG550-PVL6768-PEG550) displayed a sol-gel transition at a physiological temperature based on the test tube inverting method and rheological studies. The thermogelling copolymer demonstrated a characteristic crystalline peak for PVL block as determined by DSC and XRD analysis. In vitro release from the copolymer hydrogel matrix indicated that dexamethasone (DEX), a hydrophobic model drug, released comparatively slower than 5-fluoruracil (5-FU), a hydrophilic model drug, due to the potential partitioning of DEX into the PVL core. 5-FU in vitro release from copolymer 2 was 86% in 22 h, whereas only 14% of DEX was released in 24h. Cell viability studies confirmed that hydrogels composed of block copolymers are biocompatible. Copolymer 2 showed more than 80% relative cell viability at all concentrations, including concentrations greater than 200 fold CMC. In vivo gel formation studies indicate that gel integrity was maintained for 7 days upon subcutaneous injection into mice. These results indicate that PEG-PVL-PEG copolymers are suitable for drug delivery applications.
Product Description
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Vinyl laurate, produced by Wacker Chemie AG, Burghausen, Germany, ≥98% (GC)
Sigma-Aldrich
Vinyl laurate, ≥99.0% (GC)
Supelco
5-Fluorouracil, analytical standard
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
USP
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)