Merck
  • Natural occurrence of four Alternaria mycotoxins in tomato- and citrus-based foods in China.
Natural occurrence of four Alternaria mycotoxins in tomato- and citrus-based foods in China.
Journal of agricultural and food chemistry (2014-12-19)
Kai Zhao, Bing Shao, Dajin Yang, Fengqin Li
ABSTRACT
A total of 70 tomato-based and 86 citrus-based products collected in China were analyzed for alternariol, alternariol monomethyl ether, tentoxin, and tenuazonic acid by ultraperformance liquid chromatography–electrospray ionization–tandem mass spectrometry. No toxins were found in any fresh tomato or citrus fruit samples. Tenuazonic acid was the predominant toxin detected in all tomato ketchup (10.2–1787 μg/kg) and tomato juice samples (7.4–278 μg/kg). Alternariol was quantitated at higher level than alternariol monomethyl ether with the ratio of alternariol/alternariol monomethyl ether ranging from 0.37 to 104 in 14 alternariol-positive tomato ketchup samples. Tentoxin was detected at much lower levels in all samples analyzed. Some citrus juice samples were positive for tenuazonic acid and alternariol monomethyl ether. It is necessary to conduct a systemic surveillance of Alternaria toxins in raw and processed foods to provide the scientific basis for risk assessment of dietary exposure to these toxins in Chinese populations.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Aflatoxin B1 from Aspergillus flavus, from Aspergillus flavus
Supelco
Tenuazonic acid, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)