Merck
  • The histone methyltransferase EZH2, an oncogene common to benign and malignant parathyroid tumors.
The histone methyltransferase EZH2, an oncogene common to benign and malignant parathyroid tumors.
Endocrine-related cancer (2013-12-03)
Jessica Svedlund, Elham Barazeghi, Peter Stålberg, Per Hellman, Göran Åkerström, Peyman Björklund, Gunnar Westin
ABSTRACT
Primary hyperparathyroidism (pHPT) resulting from parathyroid tumors is a common endocrine disorder with incompletely understood etiology. In renal failure, secondary hyperparathyroidism (sHPT) occurs with multiple tumor development as a result of calcium and vitamin D regulatory disturbance. The aim of this study was to investigate a potential role of the histone 3 lysine 27 methyltransferase EZH2 in parathyroid tumorigenesis. Parathyroid tumors from patients with pHPT included adenomas and carcinomas. Hyperplastic parathyroid glands from patients with HPT secondary to uremia and normal parathyroid tissue specimens were included in this study. Quantitative RT-PCR, western blotting, bisulfite pyrosequencing, colony formation assay, and RNA interference were used. EZH2 was overexpressed in a subset of the benign and in all malignant parathyroid tumors as determined by quantitative RT-PCR and western blotting analyses. Overexpression was explained by EZH2 gene amplification in a large fraction of tumors. EZH2 depletion by RNA interference inhibited sHPT-1 parathyroid cell line proliferation as determined by tritium-thymidine incorporation and colony formation assays. EZH2 depletion also interfered with the Wnt/β-catenin signaling pathway by increased expression of growth-suppressive AXIN2, a negative regulator of β-catenin stability. Indeed, EZH2 contributed to the total level of aberrantly accumulated transcriptionally active (nonphosphoylated) β-catenin in the parathyroid tumor cells. To our knowledge EZH2 gene amplification presents the first genetic aberration common to parathyroid adenomas, secondary hyperplastic parathyroid glands, and parathyroid carcinomas. This supports the possibility of a common pathway in parathyroid tumor development.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Lrp5
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ezh2
Sigma-Aldrich
MISSION® esiRNA, targeting human EZH2
Sigma-Aldrich
ChIPAb+ EZH2, clone AC22 - ChIP Validated Antibody and Primer Set, clone AC22, from mouse
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
Sigma-Aldrich
Anti-PARP Poly ADP-ribose Polymerase-1 Antibody, serum, Chemicon®