Merck
  • In vitro degradability, bioactivity and cell responses to mesoporous magnesium silicate for the induction of bone regeneration.
In vitro degradability, bioactivity and cell responses to mesoporous magnesium silicate for the induction of bone regeneration.
Colloids and surfaces. B, Biointerfaces (2014-06-07)
Zhaoying Wu, Tingting Tang, Han Guo, Songchao Tang, Yunfei Niu, Jue Zhang, Wenjing Zhang, Rui Ma, Jiacan Su, Changsheng Liu, Jie Wei
ABSTRACT
Mesoporous magnesium silicate (m-MS) was synthesized, and the in vitro degradability, bioactivity and primary cell responses to m-MS were investigated. The results suggested that the m-MS with mesoporous channels of approximately 5nm possessed the high specific surface area of 451.0m(2)/g and a large specific pore volume of 0.41cm(3)/g compared with magnesium silicate (MS) without mesopores of 75m(2)/g and 0.21cm(3)/g, respectively. The m-MS was able to absorb a large number of water, with water absorption of 74% compared with 26% for MS. The m-MS was also degradable in a Tris-HCl solution, with a weight loss ratio of 40wt% after a 70-day immersion period. The m-MS exhibited good in vitro bioactivity, inducing apatite formation on its surfaces after soaking in simulated body fluid (SBF) at a faster rate than observed for MS. The m-MS surface clearly promoted the proliferation and differentiation of MC3T3-E1 cells, and their normal cell morphology indicated excellent cytocompatibility. This study suggested that mesoporous magnesium silicate with a high specific surface area and pore volume had suitable degradability and good bioactivity and biocompatibility, making it an excellent candidate biomaterial for the induction of bone regeneration.
Sigma-Aldrich
Magnesium nitrate hexahydrate, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Magnesium nitrate hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Magnesium nitrate hexahydrate, ACS reagent, 99%
Sigma-Aldrich
Magnesium nitrate hexahydrate, BioXtra, ≥98%
Sigma-Aldrich
Magnesium nitrate hexahydrate, puriss. p.a., ACS reagent, 98.0-102.0% (KT)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Supelco
Dimethyl sulfoxide, analytical standard