Merck
  • Repair of spinal cord injury by implantation of bFGF-incorporated HEMA-MOETACL hydrogel in rats.
Repair of spinal cord injury by implantation of bFGF-incorporated HEMA-MOETACL hydrogel in rats.
Scientific reports (2015-03-13)
Bo Chen, Jianyu He, Hao Yang, Qian Zhang, Lingling Zhang, Xian Zhang, En Xie, Cuicui Liu, Rui Zhang, Yi Wang, Linhong Huang, Dingjun Hao
ABSTRACT
There is no effective strategy for the treatment of spinal cord injury (SCI). An appropriate combination of hydrogel materials and neurotrophic factor therapy is currently thought to be a promising approach. In this study, we performed experiments to evaluate the synergic effect of implanting hydroxyl ethyl methacrylate [2-(methacryloyloxy)ethyl] trimethylammonium chloride (HEMA-MOETACL) hydrogel incorporated with basic fibroblast growth factor (bFGF) into the site of surgically induced SCI. Prior to implantation, the combined hydrogel was surrounded by an acellular vascular matrix. Sprague-Dawley rats underwent complete spinal cord transection at the T-9 level, followed by implantation of bFGF/HEMA-MOETACL 5 days after transection surgery. Our results showed that the bFGF/HEMA-MOETACL transplant provided a scaffold for the ingrowth of regenerating tissue eight weeks after implantation. Furthermore, this newly designed implant promoted both nerve tissue regeneration and functional recovery following SCI. These results indicate that HEMA-MOETACL hydrogel is a promising scaffold for intrathecal, localized and sustained delivery of bFGF to the injured spinal cord and provide evidence for the possibility that this approach may have clinical applications in the treatment of SCI.
Sigma-Aldrich
Bis[3-(triethoxysilyl)propyl] tetrasulfide, technical, ≥90% (NMR)
Pricing and availability is not currently available.