Merck
  • In vitro and in vivo evaluation under finite conditions of a transdermal oil-in-water type emulsified formulation of propiverine hydrochloride.
In vitro and in vivo evaluation under finite conditions of a transdermal oil-in-water type emulsified formulation of propiverine hydrochloride.
Biological & pharmaceutical bulletin (2014-10-03)
Rakan Matsui, Shinya Uchida, Noriyuki Namiki
ABSTRACT
A transdermal oil-in-water type emulsified formulation containing propiverine hydrochloride, used for treatment of an overactive bladder (OAB), was evaluated for in vitro skin permeation under finite conditions and in vivo transdermal absorption. Propiverine hydrochloride solubility was determined using 1,3-butyleneglycol, polyoxyethylene (2) oleylether, isostearyl alcohol, and lauryl alcohol. The solubility increased as the solubility parameter value increased. In vitro skin permeation in hairless mouse skin and in vivo transdermal absorption in rats were measured using propiverine hydrochloride dissolved in a simple solution containing these solvents. Dependent on the increase in in vitro flux, the in vivo area under the curve up to 72 h (AUC0-72) was increased. Therefore, the emulsified formulation was prepared containing these ingredients using polyoxyethylene (20) stearylether for optimization. The emulsified formulation was used to conduct in vivo single- and repeated-dose absorption studies in rats. After single-dose transdermal administration of the emulsified formulation, the AUC0-72 was equivalent to that of the simple solution. Furthermore, results using the emulsified formulation indicated an increase in AUC0-72 and significant extension of the elimination half-life, in comparison with oral administration. After repeated-dose administration, a significant minimum plasma concentration was observed compared with oral administration. These results demonstrate that the emulsified formulation is a good option for transdermal delivery of propiverine hydrochloride.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
(±)-1,3-Butanediol, anhydrous, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
(±)-1,3-Butanediol, ReagentPlus®, 99.5%
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
(±)-1,3-Butanediol, analytical standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%