Merck
  • Thermal stability of kudzu root (Pueraria Radix) isoflavones as additives to beef patties.
Thermal stability of kudzu root (Pueraria Radix) isoflavones as additives to beef patties.
Journal of food science and technology (2015-03-10)
S Kumari, J M Raines, J M Martin, J M Rodriguez
ABSTRACT
Kudzu root, Pueraria radix, extracts are a rich source of isoflavones. This study investigates the thermal stability of Pueraria radix extracts as a natural nutraceutical supplement in beef patties. The extract contained puerarin, diadzin, genistin, ononin, daidzein, glycitein, calycosin, genistein, formononetin and biochanin A; however, puerarin, daidzein and glycitein were the main components. The isoflavones concentrations in the spiked beef patties with kudzu root extracts were unaffected by cooking.
Sigma-Aldrich
Ononin, ≥99.0% (TLC)
Sigma-Aldrich
Calycosin, ≥98% (HPLC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%