Merck
  • Photo-responsive and NGR-mediated multifunctional nanostructured lipid carrier for tumor-specific therapy.
Photo-responsive and NGR-mediated multifunctional nanostructured lipid carrier for tumor-specific therapy.
Journal of pharmaceutical sciences (2015-01-30)
Yanfang Yang, Xiangyang Xie, Yang Yang, Hui Zhang, Xingguo Mei
ABSTRACT
A novel nanostructured lipid carrier (NLC) modified with photon-sensitive cell penetrating peptides (psCPP) and Asn-Gly-Arg (NGR) was designed to enhance paclitaxel (PTX)-targeted delivery and antitumor effect. The NGR moiety selectively binds to CD13-positive tumors. On other hand, the psCPP moiety enhance specific cancer cellular uptake after rapidly cleaving the two-photon excitation-responsive protective group, in this case, illumination in the presence of near-IR (NIR) light at the tumor site. The dual-modified NLC (psCPP/NGR-NLC) were prepared by emulsification method, and the concentrations of 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-polyethylene glycol 2000-psCPP (DSPE-PEG2000 -psCPP) and DSPE-PEG5000 -NGR in the NLC were chosen to be 4% and 1% (molar ratio), respectively. The mean particle size of the psCPP/NGR-NLC was about 100 nm, and the drug entrapment efficiency was more than 90%. Stability study showed that the prepared NLCs were physically and chemically stable at 2°C-8°C up to 1 month. Cellular uptake results demonstrated that the proposed psCPP/NGR-NLC had an enhancement of cancer cell recognition and specific uptake. Pharmacokinetic study showed that the prepared psCPP/NGR-NLC possessed the long-circulation characteristic with the t1/2 of 6.112 ± 0.304 h. Pharmacodynamics results confirmed that, with the aid of NIR illumination and NGR, the tumor inhibition ratio of psCPP/NGR-NLC group was significantly higher than the other PTX groups.
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Supelco
Triethylamine, analytical standard
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%