Merck
  • Synthesis of PS/PO-chimeric oligonucleotides using mixed oxathiaphospholane and phosphoramidite chemistry.
Synthesis of PS/PO-chimeric oligonucleotides using mixed oxathiaphospholane and phosphoramidite chemistry.
Organic & biomolecular chemistry (2014-11-05)
Ewa Radzikowska, Janina Baraniak
ABSTRACT
Chimeric oligonucleotides containing phosphodiester and phosphorothioate linkages have been obtained using the solid phase synthesis. The oligonucleotide parts possessing natural internucleotide phosphate bonds were assembled using commercially available nucleoside 3'-O-(2-cyanoethyl-N,N-diisopropylamino)phosphoramidites 7 whereas the phosphorothioate segment was built using nucleoside 3'-O-(2-thio-1,3,2-oxathiaphospholanes) 3. The oxidation steps, crucial for the conversion of phosphite linkages into the phosphate moieties, were conducted using tert-butylperoxy-trimethylsilane, and this reagent was not harmful to the diester phosphorothioate linkages. When P-diastereopure nucleoside 3'-O-(2-thio-1,3,2-oxathiaphospholane) monomers were employed the resulting chimeric backbone retained the P-stereoregularity of the phosphorothioate units.
Product Description
Supelco
Piperidine, analytical standard
Supelco
Decane, analytical standard
Sigma-Aldrich
Fmoc-Sar-OH, ≥98.0%
Sigma-Aldrich
1,8-Diazabicyclo[5.4.0]undec-7-ene, puriss., ≥99.0% (GC)
Sigma-Aldrich
Decane, anhydrous, ≥99%
Supelco
Decane, analytical standard
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
Tetrazole solution, suitable for DNA synthesis, filtered through a 1 μm filter, ~0.45 M in acetonitrile
Sigma-Aldrich
Ammonium citrate dibasic, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Decane, ≥95%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Decane, ReagentPlus®, ≥99%
Sigma-Aldrich
Piperidine, biotech. grade, ≥99.5%
Sigma-Aldrich
Piperidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Sigma-Aldrich
Ammonium citrate dibasic, puriss., ≥98%
Sigma-Aldrich
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98%
Sigma-Aldrich
Ammonium citrate dibasic, ACS reagent, 98%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetic anhydride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)