Merck
  • Post-harvest UV-B irradiation induces changes of phenol contents and corresponding biosynthetic gene expression in peaches and nectarines.
Post-harvest UV-B irradiation induces changes of phenol contents and corresponding biosynthetic gene expression in peaches and nectarines.
Food chemistry (2014-06-11)
Claudia Scattino, Antonella Castagna, Susanne Neugart, Helen M Chan, Monika Schreiner, Carlos H Crisosto, Pietro Tonutti, Annamaria Ranieri
ABSTRACT
In the present study the possibility of enhancing phenolic compound contents in peaches and nectarines by post-harvest irradiation with UV-B was assessed. Fruits of 'Suncrest' and 'Babygold 7' peach and 'Big Top' nectarine cultivars were irradiated with UV-B for 12 h, 24 h and 36 h. Control fruits underwent the same conditions but UV-B lamps were screened by benzophenone-treated polyethylene film. The effectiveness of the UV-B treatment in modulating the concentration of phenolic compounds and the expression of the phenylpropanoid biosynthetic genes, was genotype-dependent. 'Big Top' and 'Suncrest' fruits were affected by increasing health-promoting phenolics whereas in 'Babygold 7' phenolics decreased after UV-B irradiation. A corresponding trend was exhibited by most of tested phenylpropanoid biosynthesis genes. Based on these results UV-B irradiation can be considered a promising technique to increase the health-promoting potential of peach fruits and indirectly to ameliorate the aesthetic value due to the higher anthocyanin content.
Sigma-Aldrich
Tannic acid
Sigma-Aldrich
Tannic acid, Source: Chinese natural gall nuts
Sigma-Aldrich
Tannic acid, ACS reagent
Sigma-Aldrich
Tannic acid, puriss., powder
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)