Merck
  • Solid phase extraction of radioyttrium from irradiated strontium target using nanostructure ion imprinted polymer formed with 1-hydroxy-4-(prop-2-enyloxy)-9,10-anthraquinone.
Solid phase extraction of radioyttrium from irradiated strontium target using nanostructure ion imprinted polymer formed with 1-hydroxy-4-(prop-2-enyloxy)-9,10-anthraquinone.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2014-04-01)
P Sarabadani, M Payehghadr, M Sadeghi, Z Es'haghi, N Soltani, S Rajabifar
ABSTRACT
A new complex of 1-hydroxy-4-(prop-2-enyloxy)-9,10-anthraquinone and Y(III) was synthesized and used as the functional monomer for a novel Y(III) nanostructure ion-imprinted polymer. This polymer was used as a sorbent for fast and selective separation. The separation method was based on radioyttrium sorption from aqueous HCl solution containing strontium onto ion-imprinted polymer. Influence of various parameters such as pH, mass sorbent, nature, concentration and volume of eluent was investigated. The radioyttrium recovery yield was 99.8% and Sr content was estimated to be 3 µg mL(-1).
Product Description
Supelco
2-Methoxyethanol, analytical standard
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
2-Methoxyethanol, anhydrous, 99.8%
Supelco
2-Methoxyethanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized
Methyl methacrylate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Methoxyethanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
2-Methoxyethanol, contains 50 ppm BHT as stabilizer, ACS reagent, ≥99.3%
Sigma-Aldrich
2-Methoxyethanol, ReagentPlus®, ≥99.0%, contains 50 ppm BHT as stabilizer