Merck
  • Effects of the oral, direct factor Xa inhibitor apixaban on routine coagulation assays and anti-FXa assays.
Effects of the oral, direct factor Xa inhibitor apixaban on routine coagulation assays and anti-FXa assays.
Journal of thrombosis and haemostasis : JTH (2014-06-27)
A Hillarp, K M Gustafsson, L Faxälv, K Strandberg, F Baghaei, I Fagerberg Blixter, M Berndtsson, T L Lindahl
ABSTRACT
Apixaban is an oral direct factor Xa inhibitor developed for the prophylaxis and treatment of thromboembolic disorders. Laboratory monitoring is not necessary, but the effects on common coagulation reagents and assays constitute clinically valuable information. To investigate the effects of apixaban on commonly used coagulation methods, and to evaluate anti-FXa assays for specific determination of the drug concentration. Apixaban was added to plasma from healthy subjects in the concentration range 0-1000 μg L(-1) , and analyses were performed with different reagents for activated partial thromboplastin time (APTT), prothrombin time (PT), antithrombin, protein C, and protein S. A lupus anticoagulant assay and an APTT assay with varying phospholipid concentrations were used to study the phospholipid dependence. In general, apixaban showed fewer effects in vitro than have been shown for rivaroxaban, another direct FXa inhibitor. The concentration needed to double the APTT varied between 2200 and 4700 μg L(-1) , and the concentration needed to double the PT varied between 700 and 3900 μg L(-1) . The effects on antithrombin, protein C and protein S assays were dependent on the type of reagent. Apixaban did not cause false-positive lupus anticoagulant results. Chromogenic anti-FXa assays showed linear dose-response curves with apixaban. Therapeutic concentrations of apixaban variably affect different assay groups, and even different reagents within an assay group. The effects were much smaller than with rivaroxaban. The use of APTT and/or PT assays to screen the anticoagulant activity of apixaban cannot be recommended. A chromogenic anti-FXa assay can be used for reliable measurements of apixaban concentration.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)