Merck
  • Resveratrol is not compatible with a Fura-2-based assay for measuring intracellular Ca²⁺ signaling.
Resveratrol is not compatible with a Fura-2-based assay for measuring intracellular Ca²⁺ signaling.
Biochemical and biophysical research communications (2014-07-19)
Ram Chandra Paudel, Santeri Kiviluoto, Jan B Parys, Geert Bultynck
ABSTRACT
Fura-2 is a commonly used fluorescent Ca(2+) dye that allows an accurate determination of cytosolic Ca(2+) levels by measuring the emission obtained at 510 nm after alternating excitation at 340 nm and 380 nm (F340/F380 ratio). Previous studies, based on Fura-2 measurements, claimed that resveratrol, a polyphenol implicated in human health, triggered an acute rise in cytosolic [Ca(2+)]. In this report, we show that the spectral properties of resveratrol are not compatible with the fluorescent properties of Fura-2. Resveratrol displays a strong absorption of light at a wavelength of 340 nm and a strong emission at 510 nm upon excitation at 340 nm (F340). As a consequence, the F340 values, but not the F380 values, are increased when incubating cells with resveratrol. Consequently the F340/F380 ratio values acutely increase upon addition of resveratrol, independently of changes in cytosolic [Ca(2+)]. Yet, we show that pretreating cells with resveratrol does not affect the F340/F380 ratios of Fura-2, provided that resveratrol is washed away before fluorescence measurement. These results indicate that Fura-2 is not suitable for assessing acute effects of resveratrol on Ca(2+) signaling but that long-time effects can be assessed, provided that the resveratrol is carefully removed by appropriate wash steps.
Product Description
Supelco
Resveratrol, analytical standard
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Fura 2 pentapotassium salt, ≥80%
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Resveratrol, European Pharmacopoeia (EP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)