Merck
  • Cardiovascular risk factors among people with severe mental illness in Italy: a cross-sectional comparative study.
Cardiovascular risk factors among people with severe mental illness in Italy: a cross-sectional comparative study.
General hospital psychiatry (2014-09-14)
Massimo Clerici, Francesco Bartoli, Daniele Carretta, Cristina Crocamo, Paul Bebbington, Giuseppe Carrà
ABSTRACT
This study aimed to explore the prevalence of cardiovascular disease (CVD) risk factors in people with severe mental illness (SMI), estimating its weight on related risks in Southern Europe, an area with distinct dietary traditions. People with SMI may be more likely than general population to have high prevalence of risk factors for CVDs, due to several reasons, including poor health behaviours and psychopharmacological medications. A cross-sectional study has been conducted, comparing inpatients with and without SMI. Univariate and multivariate analyses and logistic regression models have been carried out, exploring the effect of SMI on the prevalence of CVD risk factors. Although obesity and hypertriglyceridemia were more prevalent among people with SMI, controlling for relevant confounders in multivariate analyses we could not detect any significant difference. People with SMI may not necessarily have higher CVD risk, as compared with general population. Mediterranean healthy dietary habits, universal health coverage, families providing strong support, all are possible explanations for our findings. Further research is needed, exploring different mechanisms, with more detailed biological and genetics measures.
Product Description
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Supelco
Chlorpromazine hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Sigma-Aldrich
Chlorpromazine hydrochloride, meets USP testing specifications
Chlorpromazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Chlorpromazine hydrochloride, United States Pharmacopeia (USP) Reference Standard