Merck
  • Synergistic effect of fermented rice extracts on the probiotic and laxative properties of yoghurt in rats with loperamide-induced constipation.
Synergistic effect of fermented rice extracts on the probiotic and laxative properties of yoghurt in rats with loperamide-induced constipation.
Evidence-based complementary and alternative medicine : eCAM (2014-09-13)
Jae-Suk Choi, Joo Wan Kim, Ki-Young Kim, Jong-Kwang Lee, Jae Hak Sohn, Sae-Kwang Ku
ABSTRACT
Aim. The objective was to evaluate the synergistic effects of fermented rice extracts (FRe) on the laxative and probiotic properties of yoghurt in rats with loperamide-induced constipation. Methods. After constipation induction, yoghurt containing FRe (BFRe; 0.05%, 0.1%, or 1%) was administered orally once per day for 6 days. Results. Loperamide treatment caused marked decreases in fecal pellet numbers and water content discharged, as well as in the surface mucosal thickness of the colonic lumen, intestinal charcoal transit ratio, thickness, and number of mucous-producing goblet cells in the colonic mucosa, whereas it increased the remnant fecal pellet number and the mean diameter of the colonic lumen. However, this loperamide-induced constipation was ameliorated by treatment with FRe, yoghurt single formula, or 0.05%, 0.1%, or 1% BFRe (10 mL/kg). Additionally, the viable numbers of Lactobacillus in the cecal contents and feces were markedly higher than those in constipated rats. Moreover, greater probiotic and laxative effects were detected in BFRe-treated rats than in rats treated with equivalent doses of yoghurt or FRe single formula. Conclusion. The results suggest that addition of FRe to liquid yoghurt will enhance the probiotic and beneficial laxative effects of yoghurt in the digestive tract, without causing side effects.
Product Description
Loperamide hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Loperamide hydrochloride
Supelco
Loperamide hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Loperamide hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Loperamide hydrochloride, VETRANAL®, analytical standard