Merck
  • Maresin biosynthesis and identification of maresin 2, a new anti-inflammatory and pro-resolving mediator from human macrophages.
Maresin biosynthesis and identification of maresin 2, a new anti-inflammatory and pro-resolving mediator from human macrophages.
PloS one (2014-07-19)
Bin Deng, Chin-Wei Wang, Hildur H Arnardottir, Yongsheng Li, Chien-Yee Cindy Cheng, Jesmond Dalli, Charles N Serhan
ABSTRACT
Maresins are a new family of anti-inflammatory and pro-resolving lipid mediators biosynthesized from docosahexaenoic acid (DHA) by macrophages. Here we identified a novel pro-resolving product, 13R,14S-dihydroxy-docosahexaenoic acid (13R,14S-diHDHA), produced by human macrophages. PCR mapping of 12-lipoxygenase (12-LOX) mRNA sequence in human macrophages and platelet showed that they are identical. This human 12-LOX mRNA and enzyme are expressed in monocyte-derived cell lineage, and enzyme expression levels increase with maturation to macrophages or dendritic cells. Recombinant human 12-LOX gave essentially equivalent catalytic efficiency (kcat/KM) with arachidonic acid (AA) and DHA as substrates. Lipid mediator metabololipidomics demonstrated that human macrophages produce a novel bioactive product 13,14-dihydroxy-docosahexaenoic acid in addition to maresin-1, 7R,14S-dihydroxy-4Z,8E,10E,12Z,16Z,19Z-docosahexaenoic acid (MaR1). Co-incubations with human recombinant 12-LOX and soluble epoxide hydrolase (sEH) demonstrated that biosynthesis of 13,14-dihydroxy-docosahexaenoic acid (13,14-diHDHA) involves the 13S,14S-epoxy-maresin intermediate produced from DHA by 12-LOX, followed by conversion via soluble epoxide hydrolase (sEH). This new 13,14-diHDHA displayed potent anti-inflammatory and pro-resolving actions, and at 1 ng reduced neutrophil infiltration in mouse peritonitis by ∼40% and at 10 pM enhanced human macrophage phagocytosis of zymosan by ∼90%. However, MaR1 proved more potent than the 13R,14S-diHDHA at enhancing efferocytosis with human macrophages. Taken together, the present findings demonstrate that macrophages produced a novel bioactive product identified in the maresin metabolome as 13R,14S-dihydroxy-docosahexaenoic acid, from DHA via conversion by human 12-LOX followed by sEH. Given its potent bioactions, we coined 13R,14S-diHDHA maresin 2 (MaR2).
Sigma-Aldrich
NGF-β human, from human, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Tricine, ≥99% (titration)
Sigma-Aldrich
Tricine, BioXtra, pH 4.0-6.0 (1 M in H2O), ≥99% (titration)
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Tricine, BioPerformance Certified, suitable for cell culture, ≥99% (titration)
SAFC
Tricine
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
USP
Ampicillin, United States Pharmacopeia (USP) Reference Standard
Supelco
Ampicillin, analytical standard