Skip to Content
Fechas de fin de año: Facturación de pedidos para envío en Ciudad de México, zona Metropolitana del Valle de México y Toluca hasta 13/12. Para otras regiones, hasta el 11/12. Receso de Merck Mexico del 18/12 al 02/01.
Merck
  • hMOB3 modulates MST1 apoptotic signaling and supports tumor growth in glioblastoma multiforme.
hMOB3 modulates MST1 apoptotic signaling and supports tumor growth in glioblastoma multiforme.
Cancer research (2014-05-30)
Fengyuan Tang, Lei Zhang, Gongda Xue, Debby Hynx, Yuhua Wang, Peter D Cron, Christian Hundsrucker, Alexander Hergovich, Stephan Frank, Brian A Hemmings, Debora Schmitz-Rohmer
ABSTRACT
New therapeutic targets are needed that circumvent inherent therapeutic resistance of glioblastoma multiforme (GBM). Here, we report such a candidate target in the uncharacterized adaptor protein hMOB3, which we show is upregulated in GBM. In a search for its biochemical function, we found that hMOB3 specifically interacts with MST1 kinase in response to apoptotic stimuli and cell-cell contact. Moreover, hMOB3 negatively regulated apoptotic signaling by MST1 in GBM cells by inhibiting the MST1 cleavage-based activation process. Physical interaction between hMOB3 and MST1 was essential for this process. In vivo investigations established that hMOB3 sustains GBM cell growth at high cell density and promotes tumorigenesis. Our results suggest hMOB3 as a candidate therapeutic target for the treatment of malignant gliomas.
Sigma-Aldrich
Cyclohexylamine, ReagentPlus®, ≥99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
1 Estimated to ship on February 23, 2025
MXP 1,486.00
500 g
1 Estimated to ship on February 23, 2025
MXP 4,676.00
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
1 Estimated to ship on February 23, 2025
MXP 1,486.00
500 g
1 Estimated to ship on February 23, 2025
MXP 4,676.00
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
1 Estimated to ship on February 23, 2025
MXP 1,486.00
500 g
1 Estimated to ship on February 23, 2025
MXP 4,676.00
Sigma-Aldrich
Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
1 Estimated to ship on February 23, 2025
MXP 1,486.00
500 g
1 Estimated to ship on February 23, 2025
MXP 4,676.00
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
1 Estimated to ship on February 23, 2025
MXP 1,486.00
500 g
1 Estimated to ship on February 23, 2025
MXP 4,676.00
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
1 Estimated to ship on February 23, 2025
MXP 1,486.00
500 g
1 Estimated to ship on February 23, 2025
MXP 4,676.00
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
1 Estimated to ship on February 23, 2025
MXP 1,486.00
500 g
1 Estimated to ship on February 23, 2025
MXP 4,676.00
Etoposide, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
1 Estimated to ship on February 23, 2025
MXP 1,486.00
500 g
1 Estimated to ship on February 23, 2025
MXP 4,676.00