Merck
  • Quality by design (QbD) based development and validation of an HPLC method for amiodarone hydrochloride and its impurities in the drug substance.
Quality by design (QbD) based development and validation of an HPLC method for amiodarone hydrochloride and its impurities in the drug substance.
Journal of pharmaceutical and biomedical analysis (2014-08-30)
S Karmarkar, X Yang, R Garber, A Szajkovics, M Koberda
ABSTRACT
The USP monograph describes an HPLC method for seven impurities in the amiodarone drug substance using a L1 column, 4.6mm×150mm, 5μm packing (PF listed ODS2 GL-Science, Inertsil column) at 30°C with detection at 240nm. The standard contains 0.01mg/mL of amiodarone, and USP specified impurities D and E with a resolution requirement of NLT 3.5 between peaks D and E. Impurities in a 5mg/mL sample are quantitated against the standard. Impurity A peak elutes just before peak D. We observed two problems with the method; the column lot-to-lot variability resulted in unresolved A, D, and E peaks, and peak D in the sample preparation eluted much later than that in the standard solution. Therefore, optimization experiments were conducted on the USP method following the QbD approach with Fusion AE™ software (S-Matrix Corporation). The resulting optimized conditions were within the allowable changes per USP 〈621〉. Lot-to-lot variability was negligible with the Atlantis T3 (Waters Corporation) L1 column. Peak D retention time remained constant from standard to sample. The optimized method was validated in terms of accuracy, precision, linearity, range, LOQ/LOD, specificity, robustness, equivalency to the USP method, and solution stability. The QbD based development helped in generating a design space and operating space with knowledge of all method performance characteristics and limitations and successful method robustness within the operating space.
Product Description
Supelco
Acetic acid, analytical standard
Supelco
Amiodarone hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
Sigma-Aldrich
Ammonium hydroxide solution, puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a. plus, ≥25% NH3 in H2O
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard