Merck
  • Cytokine profiles in mouse models of experimental immune thrombocytopenia reveal a lack of inflammation and differences in response to intravenous immunoglobulin depending on the mouse strain.
Cytokine profiles in mouse models of experimental immune thrombocytopenia reveal a lack of inflammation and differences in response to intravenous immunoglobulin depending on the mouse strain.
Transfusion (2014-05-16)
Danila Leontyev, Anton Neschadim, Donald R Branch
ABSTRACT
Mouse models of human immune thrombocytopenia (ITP) have been used for years to investigate the mechanism of intravenous immunoglobulin (IVIG) to ameliorate ITP; however, how closely these experimental mouse models mirror the human autoimmune inflammatory disease is unclear. The aim of this study was to assess the cytokine profiles in experimental ITP with and without IVIG treatment. We examined the production of 23 cytokines that included pro- and anti-inflammatory cytokines, in two different mouse strain models of ITP, BALB/c and C57BL/6J, with and without IVIG treatment. In contrast to human ITP, in mouse models of ITP generated by passive transfer of an alloantibody we find no evidence of inflammatory disease even when ITP is robust, suggesting that while these models capture the innate response phase of the disease, they may not be capturing the adaptive mechanisms of autoimmune initiation of the disorder. Regardless of the mouse strain examined, interleukin (IL)-1α, -2, -6, -17, and -23; granulocyte-macrophage-colony-stimulating factor (GM-CSF); monocyte chemoattractant protein (MCP)-1; macrophage inflammatory protein (MIP)-1β; RANTES; tumor necrosis factor (TNF)-α; and interferon-γ remain at negligible levels after ITP induction. IVIG treatment in the absence of ITP induced significant levels of IL-4, -10, -11, -17, and -23; GM-CSF; MCP-1; and TNF-α in BALB/c mice, but only IL-11 was elevated in C57BL/6J mice. In response to IVIG treatment of ITP, both strains produced IL-4, -10, -11, and -23; GM-CSF; MCP-1; MIP-1β; and TNF-α; however, only BALB/c exhibited increased MCP-3 and RANTES. IL-11 levels were the highest of any cytokine after IVIG administration, given either alone or as treatment of ITP. We conclude that mouse models for human ITP do not capture the full range of autoimmune inflammatory mechanisms of this disease. Furthermore, cytokine profiles differ in response to IVIG depending on the mouse strain used.
Sigma-Aldrich
Mouse Il33 / Interleukin-33 ELISA Kit
Sigma-Aldrich
Mouse IL-11 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human IL-11 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human IL-33 ELISA Kit, for serum, plasma, cell culture supernatant and urine