Merck
  • In vitro and in vivo activities of three oxazolidinones against nonreplicating Mycobacterium tuberculosis.
In vitro and in vivo activities of three oxazolidinones against nonreplicating Mycobacterium tuberculosis.
Antimicrobial agents and chemotherapy (2014-03-26)
Ming Zhang, Claudia Sala, Neeraj Dhar, Anthony Vocat, Vasan K Sambandamurthy, Sreevalli Sharma, Gwendolyn Marriner, V Balasubramanian, Stewart T Cole
ABSTRACT
Oxazolidinones represent a new class of antituberculosis drugs that exert their function by inhibiting protein synthesis. Here, we compared the activities of three oxazolidinones, linezolid, PNU-100480, and AZD5847, against latent tuberculosis using a simple model employing the streptomycin-starved Mycobacterium tuberculosis strain 18b. The in vitro drug susceptibility results showed that the three oxazolidinones had a bacteriostatic effect against actively growing bacilli but potent bactericidal activity against nonreplicating cells. In the murine model of latent infection with M. tuberculosis 18b, the efficacy of the three compounds varied greatly. Indeed, AZD5847 or its prodrug exhibited no activity or only modest activity, respectively, after 2 months of treatment, whereas both linezolid and PNU-100480 were effective against latent bacilli in mice and showed promising outcomes in combination therapy with rifampin. Moreover, the potency of PNU-100480 was significantly greater than that of linezolid, making it an attractive drug candidate in the development of new combination therapies for latent tuberculosis.
Product Description
Supelco
Chloramphenicol, VETRANAL®, analytical standard
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard
USP
Clarithromycin Identity, United States Pharmacopeia (USP) Reference Standard
Isoniazid, European Pharmacopoeia (EP) Reference Standard
Amikacin, European Pharmacopoeia (EP) Reference Standard
USP
Amikacin, United States Pharmacopeia (USP) Reference Standard
Supelco
Clarithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Clarithromycin, ≥95% (HPLC)
Sigma-Aldrich
Clarithromycin, 96.0-102.0% (HPLC)
Sigma-Aldrich
Fusidic acid
Clarithromycin, European Pharmacopoeia (EP) Reference Standard
Clarithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Fusidic acid, European Pharmacopoeia (EP) Reference Standard