Merck
  • Dietary L-methionine supplementation mitigates gamma-radiation induced global DNA hypomethylation: enhanced metabolic flux towards S-adenosyl-L-methionine (SAM) biosynthesis increases genomic methylation potential.
Dietary L-methionine supplementation mitigates gamma-radiation induced global DNA hypomethylation: enhanced metabolic flux towards S-adenosyl-L-methionine (SAM) biosynthesis increases genomic methylation potential.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-04-12)
Vipen Batra, Poonam Verma
ABSTRACT
The objective of this study was to examine the effect of (60)Co-gamma (γ) radiation on modulation of genomic DNA methylation, if any, of mice maintained (6 weeks) on normal control diet (NCD) and L-methionine supplemented diet (MSD). To elucidate the possible underlying mechanism(s), we exposed the animals to γ-radiation (2, 3 and 4 Gy) and investigated the profile of downstream metabolites and enzymes involved in S-adenosyl-L-methionine (SAM) generation. Liver samples were also subjected to histopathological examinations. Compared to NCD fed and irradiated animals, hepatic folate, choline and L-methionine levels decreased moderately, while hepatic SAM levels increased in MSD fed and irradiated animals. Under these conditions, a marked modulation of methionine adenosyltransferase (MAT) and L-methionine synthase (MSase) activities was observed. Concomitant with increase in liver SAM pool, increased DNA methyltransferase (dnmt) activity in MSD fed mice indicated enhanced metabolic flux towards DNA methylation. Further results showed that genomic DNA methylation and 5-methyl-2'-deoxy cytidine residues were maintained at normal levels in MSD fed and irradiated mice compared to NCD fed and irradiated animals. In conclusion, our results suggest that increasing supply of preformed methyl groups, via dietary L-methionine supplementation might significantly increase methylation potential of radiation stress compromised DNA methylation cycle.
Sigma-Aldrich
Choline bitartrate
Sigma-Aldrich
L-Homocysteine, ≥98.0% (NT)
Supelco
Choline Bitartrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis