Merck
  • Miro1 regulates intercellular mitochondrial transport & enhances mesenchymal stem cell rescue efficacy.
Miro1 regulates intercellular mitochondrial transport & enhances mesenchymal stem cell rescue efficacy.
The EMBO journal (2014-01-17)
Tanveer Ahmad, Shravani Mukherjee, Bijay Pattnaik, Manish Kumar, Suchita Singh, Manish Kumar, Rakhshinda Rehman, Brijendra K Tiwari, Kumar A Jha, Amruta P Barhanpurkar, Mohan R Wani, Soumya S Roy, Ulaganathan Mabalirajan, Balaram Ghosh, Anurag Agrawal
ABSTRACT
There is emerging evidence that stem cells can rejuvenate damaged cells by mitochondrial transfer. Earlier studies show that epithelial mitochondrial dysfunction is critical in asthma pathogenesis. Here we show for the first time that Miro1, a mitochondrial Rho-GTPase, regulates intercellular mitochondrial movement from mesenchymal stem cells (MSC) to epithelial cells (EC). We demonstrate that overexpression of Miro1 in MSC (MSCmiro(Hi)) leads to enhanced mitochondrial transfer and rescue of epithelial injury, while Miro1 knockdown (MSCmiro(Lo)) leads to loss of efficacy. Treatment with MSCmiro(Hi) was associated with greater therapeutic efficacy, when compared to control MSC, in mouse models of rotenone (Rot) induced airway injury and allergic airway inflammation (AAI). Notably, airway hyperresponsiveness and remodeling were reversed by MSCmiro(Hi) in three separate allergen-induced asthma models. In a human in vitro system, MSCmiro(Hi) reversed mitochondrial dysfunction in bronchial epithelial cells treated with pro-inflammatory supernatant of IL-13-induced macrophages. Anti-inflammatory MSC products like NO, TGF-β, IL-10 and PGE2, were unchanged by Miro1 overexpression, excluding non-specific paracrine effects. In summary, Miro1 overexpression leads to increased stem cell repair.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rhot1
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Rotenone, ≥95%
Supelco
Rotenone, PESTANAL®, analytical standard
Sigma-Aldrich
MISSION® esiRNA, targeting human RHOT1
Dinoprostone, European Pharmacopoeia (EP) Reference Standard