Merck
  • Over-expression of telomere binding factors (TRF1 & TRF2) in renal cell carcinoma and their inhibition by using SiRNA induce apoptosis, reduce cell proliferation and migration invitro.
Over-expression of telomere binding factors (TRF1 & TRF2) in renal cell carcinoma and their inhibition by using SiRNA induce apoptosis, reduce cell proliferation and migration invitro.
PloS one (2015-03-03)
Deeksha Pal, Ujjawal Sharma, Shrawan Kumar Singh, Nandita Kakkar, Rajendra Prasad
ABSTRACT
Telomere binding factors viz. TRF1 and TRF2 are a part of sheltrin complex that are present exclusively at the ends of chromosomes. These factors play an important role in maintaining chromosomal integrity at the ends. However, their status and role are not clear in renal cell carcinoma (RCC). Therefore, the present study was conducted to evaluate TRF1 and TRF2 expressions in RCC tissues. Further, the role of these factors involved in tumorigenesis was elucidated by gene silencing using siRNA in RCC cell line (A498). The present study documented a significant over-expression of TRF1 (P = 0.005) and TRF2 (P = 0.0048) mRNAs by real time PCR in RCC tissues as compared with adjacent normal kidney tissues. Immunohistochemistry studies also revealed higher expression of TRF1 and TRF2 proteins in RCC. Moreover, TRF1 or TRF2 gene silencing using siRNA showed marked reduction in proliferation of RCC cells (P = 0.000). Further, significantly induced cell cycle arrest (P = 0.000) and apoptosis of RCC cells (P = 0.000) was documented upon TRF1 or TRF2 gene silencing. Henceforth, the results deduce that TRF1 or TRF2 inhibitions play an important role in the induction of apoptosis in A498 cells, which may serve as a potential therapeutic target in RCC.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
MISSION® esiRNA, targeting human TERF2
Sigma-Aldrich
MISSION® siRNA Universal Negative Control #1