Merck
  • Seasonal and diurnal variation of organic ultraviolet filters from personal care products used along the Japanese coast.
Seasonal and diurnal variation of organic ultraviolet filters from personal care products used along the Japanese coast.
Archives of environmental contamination and toxicology (2014-12-07)
Kenshi Sankoda, Kotaro Murata, Mai Tanihata, Kengo Suzuki, Kei Nomiyama, Ryota Shinohara
ABSTRACT
This study aimed to investigate the behavior of organic ultraviolet (UV) filters released by recreational activities along the Japanese coastline. Seasonal variations of organic UV filters in seawater were investigated at four different recreational beaches (Mogushi, Wakamiya, Tsurugahama, and Otachimisaki beaches) in both summer (July through August) and winter (December). Moreover, short time scale diurnal changes were monitored at Otachimisaki beach in summer. Of the four sunscreen agents tested in this study, two agents-2-ethylhexyl-4-methoxycinnamate (EHMC) and 2-ethylhexyl salicylate (EHS)-were detected in all samples, whereas octyl-dimethyl-p-aminobenzonic acid and 3-(4-methylbenzylidene)-camphor were lower than detection limits. In particular, EHMC, one of the most popular organic UV filters, was dominant. The highest concentration of EHMC was observed at 1,080 ng L(-1), a level that exceeds those of previous studies. Both EHMC and EHS concentrations showed significant (p < 0.05) seasonal variations with advancing summer suggesting direct input from recreational activities. The subsequent examination showed short time scale diurnal changes of organic UV filters on the beach. The results showed that diurnal changes in EHMC concentrations were correlated to the number of bathers. EHMC concentrations increased during the afternoon and decreased during the night, although complete attenuation during the night did not occur. EHMC persists along the coast due to low mobility and may persist the next day. This is the first study to show the natural attenuation behavior of organic UV filters along recreational beaches.
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Supelco
Hexane, analytical standard
Sigma-Aldrich
Phenanthrene-d10, 98 atom % D, 98% (CP)
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Aminobenzoic acid, ReagentPlus®, 99%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Camphor, 96%
Sigma-Aldrich
(±)-Camphor, meets analytical specification of Ph. Eur., BP, ≥95% (GC), racemic
Supelco
(−)-Camphor, analytical standard
Sigma-Aldrich
(±)-Camphor, purum, synthetic, ≥95.0% (GC)
Sigma-Aldrich
Chrysene-d12, 98 atom % D
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
4-Aminobenzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
4-Aminobenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
(±)-Camphor, ≥95.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
USP
Camphor, United States Pharmacopeia (USP) Reference Standard
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%