Merck
  • Tertiary-amine functionalized polyplexes enhanced cellular uptake and prolonged gene expression.
Tertiary-amine functionalized polyplexes enhanced cellular uptake and prolonged gene expression.
PloS one (2014-05-16)
Chia-Wen Lo, Yung Chang, Jyun-Lin Lee, Wei-Bor Tsai, Wen-Shiang Chen
ABSTRACT
Ultrasound (US) has been found to facilitate the transport of DNA across cell membranes. However, the transfection efficiency is generally low, and the expression duration of the transfected gene is brief. In this study, a tertiary polycation, Poly(2-(dimethylamino) ethyl methacrylate) (PDMAEMA), was used as a carrier for US-mediated gene transfection. Its in-vitro and in-vivo effects on the transfection efficiency and the expression duration were evaluated. A mixture of pCI-neo-luc and PDMAEMA was transfected to cultured cells or mouse muscle by exposure to 1-MHz pulse US. A strong expression of luciferase was found 10 days after the transfection in vitro regardless of US exposure. However, effective transfection only occurred in the US groups in vivo. The transfection ability depended on the weight ratio of PDMAEMA to DNA, and was different for the in-vitro and in-vivo conditions. Lower weight ratios, e.g., 0.25, exhibited better in-vivo expression for at least 45 days.
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
2-(Dimethylamino)ethyl methacrylate, contains 700-1000 ppm monomethyl ether hydroquinone as inhibitor, 98%
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Fluorescein, European Pharmacopoeia (EP) Reference Standard
2-(Dimethylamino)ethyl methacrylate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%
Sigma-Aldrich
Ammonium persulfate, reagent grade, 98%
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Amphotericin B, European Pharmacopoeia (EP) Reference Standard