Merck
  • Silver wire amplifies the signaling mechanism for IL-1beta production more than silver submicroparticles in human monocytic THP-1 cells.
Silver wire amplifies the signaling mechanism for IL-1beta production more than silver submicroparticles in human monocytic THP-1 cells.
PloS one (2014-11-15)
Hye Jin Jung, Pyo June Pak, Sung Hyo Park, Jae Eun Ju, Joong-Su Kim, Hoi-Seon Lee, Namhyun Chung
ABSTRACT
Silver materials have been widely used in diverse fields. However, their toxicity and their mechanism, especially in different forms, have not been studied sufficiently. Thus, cytotoxicity, apoptosis, and interleukin-1beta (IL-1β) production were investigated using macrophage-like THP-1 cells in the presence of Ag microparticles (AgMPs, 2.7 µm), Ag submicroparticles (AgSMPs, 150 nm), and Ag wires (AgWs, 274 nm×5.3 µm). The levels of cytotoxicity, apoptosis, and IL-1β production by AgWs were higher than those by the other two AgSMPs and AgMPs. This trend was also observed with each step of the signaling mechanism for IL-1β production, which is a single pathway affiliated with ROS generation or lysosomal rupture or both, cathepsin B, caspase-1 (NALP3 inflammasome), and finally IL-1β production in THP-1 cells. All these results suggest that, for development of safe and effective silver materials, the shape or form of silver materials should be considered, especially for macrophage cell lines because epithelial cell lines are not overly sensitive to silver materials.
Product Description
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3-tert-Butyl-4-hydroxyanisole, ≥98% (sum of isomers, GC), ≤10% 2-BHA basis (GC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Guaiacol, oxidation indicator
Guaiacol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Supelco
3-tert-Butyl-4-hydroxyanisole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Propidium iodide solution