Merck
  • Co-extruded solid solutions as immediate release fixed-dose combinations.
Co-extruded solid solutions as immediate release fixed-dose combinations.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-07-11)
L Dierickx, B Van Snick, T Monteyne, T De Beer, J P Remon, C Vervaet
ABSTRACT
The aim of this study was to develop by means of co-extrusion a multilayer fixed-dose combination solid dosage form for oral application characterized by immediate release for both layers, the layers containing different drugs with different water-solubility. In this study polymers were selected which can be combined in a co-extruded dosage form. Several polymers were screened on the basis of their processability via hot-melt extrusion, macroscopic properties, acetylsalicylic acid (ASA) decomposition and in vitro drug release. ASA and fenofibrate (FF) were incorporated as hydrophilic and hydrophobic model drugs, respectively. Based on the polymer screening experiments Kollidon® PF 12 and Kollidon® VA 64 were identified as useful ASA carriers (core), while Soluplus®, Kollidon® VA 64 and Kollidon® 30 were applicable as FF carriers (coat). The combination of Kollidon® 30 (coat) with Kollidon® PF 12 or Kollidon® VA 64 (core) failed in terms of processability via co-extrusion. All other combinations (containing 20% ASA in the core and 20% FF in the coat) were successfully co-extruded (diameter core: 2mm/thickness coat: 1mm). All formulations showed good adhesion between core and coat. ASA release from the core was complete within 15-30 min (Kollidon® PF 12) or 30-60 min (Kollidon® VA 64), while FF release was complete within 20-30 min (Kollidon® VA 64) or 60 min (Soluplus®). Differential scanning calorimetry (DSC) and X-ray diffraction (XRD) revealed that both drugs were molecularly dispersed in the carriers. Raman mapping exposed very little intermigration of both drugs at the interface. Fixed-dose combinations with good in vitro performance were successfully developed by means of co-extrusion, both layers providing immediate release.
Product Description
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium fluoride, BioUltra, ≥99.5% (F)
Sigma-Aldrich
Potassium fluoride, ≥99.97% trace metals basis
Sigma-Aldrich
Potassium fluoride, anhydrous, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard