Merck
  • Effects of buprenorphine on behavioral tests for antidepressant and anxiolytic drugs in mice.
Effects of buprenorphine on behavioral tests for antidepressant and anxiolytic drugs in mice.
Psychopharmacology (2014-09-03)
Edgardo Falcon, Kaitlyn Maier, Shivon A Robinson, Tiffany E Hill-Smith, Irwin Lucki
ABSTRACT
Buprenorphine (BPN) has been shown to rapidly improve mood in treatment-resistant depressed patients in small clinical studies. However, BPN's effects in preclinical tests for mood and antidepressant efficacy are largely unexplored. The current study examined the effects of BPN in the forced swim test (FST) and novelty-induced hypophagia (NIH) test as measures of antidepressant and anxiolytic-like effects in C57BL/6 J mice. Microdialysis was used to measure whether BPN engaged kappa-opioid receptor (KORs) in the nucleus accumbens shell (NAcSh) at a behaviorally active dose (0.25 mg/kg). BPN was tested in the FST at both 30 min and 24 h post-administration. Also measured in the FST at 24 h post-administration were the KOR antagonist norbinaltorphimine (nor-BNI), the MOR agonist morphine and the reference antidepressant desipramine. The anxiolytic effects of BPN were examined in the NIH test 24 h after treatment. The effects of acute injection of BPN and the KOR agonist U50,488 were measured on extracellular dopamine (DA) levels in the NAcSh. BPN produced significant reductions in FST immobility without changing locomotor activity and reduced approach latencies in the novel environment of the NIH test when tested 24 h after treatment. Repeated daily BPN injections for 6 days did not produce tolerance to these behavioral effects. nor-BNI produced a similar antidepressant-like response in the FST 24 h post-injection but morphine and desipramine were ineffective. BPN (0.25 mg/kg) did not alter DA levels when given alone but prevented the KOR agonist U50,488 from reducing DA levels. Acute and subchronic treatment with BPN produced antidepressant and anxiolytic-like responses in mice at doses that engage KORs. These studies support the clinical evidence that BPN may be a novel rapid-acting antidepressant medication and provides rodent models for investigating associated neurochemical mechanisms.
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, absolute, over molecular sieve (H2O ≤0.04%), ≥99.5% (GC)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, reagent grade
Supelco
Desipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, ≥99.0% (GC)
Sigma-Aldrich
Desipramine hydrochloride, ≥98% (TLC), powder
USP
Desipramine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Desipramine hydrochloride, European Pharmacopoeia (EP) Reference Standard