Merck
  • Methyl gallate from Galla rhois successfully controls clinical isolates of Salmonella infection in both in vitro and in vivo systems.
Methyl gallate from Galla rhois successfully controls clinical isolates of Salmonella infection in both in vitro and in vivo systems.
PloS one (2014-07-23)
Jang-Gi Choi, Su-Hyun Mun, Harendra S Chahar, Preeti Bharaj, Ok-Hwa Kang, Se-Gun Kim, Dong-Won Shin, Dong-Yeul Kwon
ABSTRACT
Galla rhois is a commonly used traditional medicine for the treatment of pathogenic bacteria in Korea as well as in other parts of Asia. Methyl gallate (MG), a major component of Galla Rhois, exhibits strong antibacterial activity, but its mechanism of action against Salmonella spp. is unclear. In the present study, we investigated the antibacterial actions of MG against Salmonella. The antibacterial activity determined by broth dilution method indicated that the antibacterial activity of MG against Salmonella strains ranged from 3.9 to 125 µg/ml. In vitro bacterial viability test indicated that MG significantly decreased the viability of Salmonella over 40% when combined with ATPase inhibitors. The time-kill curves showed that a combined MG and ATPase inhibitors (DCCD and NaN3) treatment reduced the bacterial counts dramatically after 24 h. Oral administration of MG showed a strong anti-bacterial activity against WS-5 infected BALB/c mice. In contrast to the untreated Salmonella infected control animals, MG treated groups showed no clinical symptoms of the disease, such as lethargy and liver damage. It was observed that MG treatment significantly increased the survival of animals from Salmonella infection, while in untreated groups all animal succumbed to disease by the sixth day post infection. Thus, the present study demonstrates the therapeutic ability of MG against Salmonella infections.
Product Description
USP
Amoxicillin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Millipore
Ticarcillin Supplement, suitable for microbiology
Norfloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sulfafurazole, European Pharmacopoeia (EP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Supelco
Nalidixic acid, analytical standard
Supelco
Ampicillin, analytical standard
Norfloxacin for peak identification, European Pharmacopoeia (EP) Reference Standard
Norfloxacin, European Pharmacopoeia (EP) Reference Standard
Sulfamethoxazole, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Sulfamethoxazole, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
Sulfamethoxazole, VETRANAL®, analytical standard
Supelco
Sulfisoxazole, VETRANAL®, analytical standard
Supelco
Chloramphenicol, VETRANAL®, analytical standard
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)