Merck
Identification and further development of potent TBK1 inhibitors.
ACS chemical biology (2014-12-30)
André Richters, Debjit Basu, Julian Engel, Meryem S Ercanoglu, Hyatt Balke-Want, Roberta Tesch, Roman K Thomas, Daniel Rauh
ABSTRACT
The cytosolic Ser/Thr kinase TBK1 was discovered to be an essential element in the mediation of signals that lead to tumor migration and progression. These findings meet the need for the identification of novel tool compounds and potential therapeutics to gain deeper insights into TBK1 related signaling and its relevance in tumor progression. Herein, we undertake the activity-based screening for unique inhibitors of TBK1 and their subsequent optimization. Initial screening approaches identified a selection of TBK1 inhibitors that were optimized using methods of medicinal chemistry. Variations of the structural characteristics of a representative 2,4,6-substituted pyrimidine scaffold resulted in improved potency. Prospective use as tool compounds or basic contributions to drug design approaches are anticipated for our improved small molecules.
Sigma-Aldrich
Autophinib, ≥98% (HPLC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
Dimethyl sulfoxide, analytical standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
TBK1 Protein, active, 10 µg, Active, N-terminal His6-tagged, recombinant, full length, human TBK1, for use in Kinase Assays.
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%