Merck
  • JPC-2997, a new aminomethylphenol with high in vitro and in vivo antimalarial activities against blood stages of Plasmodium.
JPC-2997, a new aminomethylphenol with high in vitro and in vivo antimalarial activities against blood stages of Plasmodium.
Antimicrobial agents and chemotherapy (2014-10-22)
Geoffrey W Birrell, Marina Chavchich, Arba L Ager, Hong-Ming Shieh, Gavin D Heffernan, Wenyi Zhao, Peter E Krasucki, Kurt W Saionz, Jacek Terpinski, Guy A Schiehser, Laura R Jacobus, G Dennis Shanks, David P Jacobus, Michael D Edstein
ABSTRACT
4-(tert-Butyl)-2-((tert-butylamino)methyl)-6-(6-(trifluoromethyl)pyridin-3-yl)-phenol (JPC-2997) is a new aminomethylphenol compound that is highly active in vitro against the chloroquine-sensitive D6, the chloroquine-resistant W2, and the multidrug-resistant TM90-C2B Plasmodium falciparum lines, with 50% inhibitory concentrations (IC50s) ranging from 7 nM to 34 nM. JPC-2997 is >2,500 times less cytotoxic (IC50s > 35 μM) to human (HepG2 and HEK293) and rodent (BHK) cell lines than the D6 parasite line. In comparison to the chemically related WR-194,965, a drug that had advanced to clinical studies, JPC-2997 was 2-fold more active in vitro against P. falciparum lines and 3-fold less cytotoxic. The compound possesses potent in vivo suppression activity against Plasmodium berghei, with a 50% effective dose (ED50) of 0.5 mg/kg of body weight/day following oral dosing in the Peters 4-day test. The radical curative dose of JPC-2997 was remarkably low, at a total dose of 24 mg/kg, using the modified Thompson test. JPC-2997 was effective in curing three Aotus monkeys infected with a chloroquine- and pyrimethamine-resistant strain of Plasmodium vivax at a dose of 20 mg/kg daily for 3 days. At the doses administered, JPC-2997 appeared to be well tolerated in mice and monkeys. Preliminary studies of JPC-2997 in mice show linear pharmacokinetics over the range 2.5 to 40 mg/kg, a low clearance of 0.22 liters/h/kg, a volume of distribution of 15.6 liters/kg, and an elimination half-life of 49.8 h. The high in vivo potency data and lengthy elimination half-life of JPC-2997 suggest that it is worthy of further preclinical assessment as a partner drug.
Sigma-Aldrich
Mefloquine hydrochloride, ≥98% (HPLC), powder
Mefloquine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Atovaquone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Atovaquone, ≥98% (HPLC)