Merck
  • Preservation of volumetric bone density and geometry in trans women during cross-sex hormonal therapy: a prospective observational study.
Preservation of volumetric bone density and geometry in trans women during cross-sex hormonal therapy: a prospective observational study.
Osteoporosis international : a journal established as result of cooperation between the European Foundation for Osteoporosis and the National Osteoporosis Foundation of the USA (2014-11-08)
E Van Caenegem, K Wierckx, Y Taes, T Schreiner, S Vandewalle, K Toye, J-M Kaufman, G T'Sjoen
ABSTRACT
Although trans women before the start of hormonal therapy have a less bone and muscle mass compared with control men, their bone mass and geometry are preserved during the first 2 years of hormonal therapy, despite of substantial muscle loss, illustrating the major role of estrogen in the male skeleton. The aim of this study is to examine the evolution of areal and volumetric bone density, geometry, and turnover in trans women undergoing sex steroid changes, during the first 2 years of hormonal therapy. In a prospective observational study, we examined 49 trans women (male-to-female) before and after 1 and 2 years of cross-sex hormonal therapy (CSH) in comparison with 49 age-matched control men measuring grip strength (hand dynamometer), areal bone mineral density (aBMD), and total body fat and lean mass using dual X-ray absorptiometry (DXA), bone geometry and volumetric bone mineral density, regional fat, and muscle area at the forearm and calf using peripheral quantitative computed tomography. Standardized treatment regimens were used with oral estradiol valerate, 4 mg daily (or transdermal 17-β estradiol 100 μg/24 h for patients >45 years old), both combined with oral cyproterone acetate 50 mg daily. Prior to CSH, trans women had lower aBMD at all measured sites (all p < 0.001), smaller cortical bone size (all p < 0.05), and lower muscle mass and strength and lean body mass (all p < 0.05) compared with control men. During CSH, muscle mass and strength decreased and all measures of fat mass increased (all p < 0.001). The aBMD increased at the femoral neck, radius, lumbar spine, and total body; cortical and trabecular bone remained stable and bone turnover markers decreased (all p < 0.05). Although trans women, before CSH, have a lower aBMD and cortical bone size compared with control men, their skeletal status is well preserved during CSH treatment, despite of substantial muscle loss.
Sigma-Aldrich
Estrone, ≥99%
Estrone, European Pharmacopoeia (EP) Reference Standard
USP
Estrone, United States Pharmacopeia (USP) Reference Standard
Supelco
Estrone, Pharmaceutical Secondary Standard; Certified Reference Material
Cyproterone acetate for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-Estradiol 17-valerate, ≥98%
Sigma-Aldrich
Cyproterone acetate, ≥98%
Sigma-Aldrich
Estradiol, meets USP testing specifications
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Cyproterone acetate, European Pharmacopoeia (EP) Reference Standard
Estradiol valerate, European Pharmacopoeia (EP) Reference Standard