Merck
  • Rapid evaluation and comparison of natural products and antioxidant activity in calendula, feverfew, and German chamomile extracts.
Rapid evaluation and comparison of natural products and antioxidant activity in calendula, feverfew, and German chamomile extracts.
Journal of chromatography. A (2015-02-11)
Snezana Agatonovic-Kustrin, Davoud Babazadeh Ortakand, David W Morton, Ahmad P Yusof
ABSTRACT
The present study describes a simple high performance thin layer chromatographic (HPTLC) method for the simultaneous quantification of apigenin, chamazulene, bisabolol and the use of DPPH free radical as a post-chromatographic derivatization agent to compare the free radical scavenging activities of these components in leaf and flower head extracts from feverfew, German chamomile and marigold from the Asteraceae family. Feverfew (Tanacetum parthenium) leaves have been traditionally used in the treatment of migraine with parthenolide being the main bioactive compound. However, due to similar flowers, feverfew is sometimes mistaken for the German chamomile (Matricaria recutita). Bisabolol and chamazulene are the main components in chamomile essential oil. Marigold (Calendula officinalis) was included in the study for comparison, as it belongs to the same family. Parthenolide was found to be present in all leaf extracts but was not detected in calendula flower extract. Chamazulene and bisabolol were found to be present in higher concentrations in chamomile and Calendula flowers. Apigenin was detected and quantified only in chamomile extracts (highest concentration in flower head extracts). Antioxidant activity in sample extracts was compared by superimposing the chromatograms obtained after post-chromatographic derivatization with DPPH and post-chromatographic derivatization with anisaldehyde. It was found that extracts from chamomile flower heads and leaves have the most prominent antioxidant activity, with bisabolol and chamazulene being the most effective antioxidants.
Sigma-Aldrich
Methanol, NMR reference standard
Rutin trihydrate, primary reference standard
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Supelco
Rutin trihydrate, analytical standard
Supelco
Acetic acid, analytical standard
Supelco
Apigenin, analytical standard
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Acetone, analytical standard
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Parthenolide, ≥98% (HPLC)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%