Merck
  • A new SiF-Dipropargyl glycerol scaffold as a versatile prosthetic group to design dimeric radioligands: synthesis of the [(18) F]BMPPSiF tracer to image serotonin receptors.
A new SiF-Dipropargyl glycerol scaffold as a versatile prosthetic group to design dimeric radioligands: synthesis of the [(18) F]BMPPSiF tracer to image serotonin receptors.
ChemMedChem (2014-01-01)
Puja Panwar Hazari, Jurgen Schulz, Delphine Vimont, Nidhi Chadha, Michele Allard, Magali Szlosek-Pinaud, Eric Fouquet, Anil Kumar Mishra
ABSTRACT
A novel SiX-dipropargyl glycerol scaffold (X: H, F, or (18) F) was developed as a versatile prosthetic group that provides technical advantages for the preparation of dimeric radioligands based on silicon fluoride acceptor pre- or post-labeling with fluorine-18. Rapid conjugation with the prosthetic group takes place in microwave-assisted click conjugation under mild conditions. Thus, a bivalent homodimeric SiX-dipropargyl glycerol derivatized radioligand, [(18) F]BMPPSiF, with enhanced affinity was developed by using click conjugation. High uptake of the radioligand was demonstrated in 5-HT1A receptor-rich regions in the brain with positron emission tomography. Molecular docking studies (rigid protein-flexible ligand) of BMPPSiF and known antagonists (WAY-100635, MPPF, and MefWAY) with monomeric, dimeric, and multimeric 5-HT1A receptor models were performed, with the highest G score obtained for docked BMPPSiF: -6.766 as compared with all three antagonists on the monomeric model. Multimeric induced-fit docking was also performed to visualize the comparable mode of binding under in vivo conditions, and a notably improved G score of -8.455 was observed for BMPPSiF. These data directly correlate the high binding potential of BMPPSiF with the bivalent binding mode obtained in the biological studies. The present study warrants wide application of the SiX-dipropargyl glycerol prosthetic group in the development of ligands for imaging with enhanced affinity markers for specific targeting based on peptides, nucleosides, and lipids.
Product Description
Paroxetine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Paroxetine hydrochloride hemihydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Spiperone, solid
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Paroxetine hydrochloride hemihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Paroxetine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ketanserin (+)-tartrate salt, ≥97%, solid