Merck
  • Neuroprotective effect of diphenyl diselenide in a experimental stroke model: maintenance of redox system in mitochondria of brain regions.
Neuroprotective effect of diphenyl diselenide in a experimental stroke model: maintenance of redox system in mitochondria of brain regions.
Neurotoxicity research (2014-03-13)
Fernando Dobrachinski, Michele Hinerasky da Silva, Cíntia Letícia Cardias Tassi, Nélson Rodrigues de Carvalho, Glaecir Roseni Mundstock Dias, Ronaldo Medeiros Golombieski, Elgion Lúcio da Silva Loreto, João Batista Teixeira da Rocha, Michele Rechia Fighera, Félix Alexandre Antunes Soares
ABSTRACT
Acute stroke is a major risk for morbidity and mortality in aging population. Mitochondrion has been the focus of a wide stroke-related research. This study investigated if treatment or pre-treatment with diphenyl diselenide (PhSe)2 can prevent mitochondrial damage in cerebral structures of rats induced by an ischemia and reperfusion (I/R) model. Adult male Wistar rats were assigned into five experimental groups: sham operation, ischemia/reperfusion, pre-treated + I/R, treated + I/R, and Sham + (PhSe)2. Neurological score showed the damage caused by I/R, which was partially prevented by (PhSe)2. Moreover, mitochondria of hippocampus and cortex were impaired by I/R through an increase of reactive oxygen species production, mitochondrial membrane potential (ΔΨm) and electrons flow alteration, activity of complex I deregulation as well as mitochondrial swelling. However, the ischemic damage did not induce an increase in pro-apoptotic proteins expression, but demonstrated an enhanced expression of Hsp70. The mitochondrial redox state was also altered (GSH/GSSG ratio, MnSOD, and GPx activities). Our results revealed that all treatments with (PhSe)2 significantly reduced the mitochondrial damage induced by I/R. These findings suggest that neuroprotective properties of (PhSe)2 may be attributed to the maintenance of mitochondrial redox balance.
Sigma-Aldrich
Phthaldialdehyde, for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Adrenaline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium 2,6-dichloroindophenolate hydrate, ACS reagent
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diphenyl diselenide, 98%
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Diphenyl diselenide, purum, ≥97.0% (GC)
Sigma-Aldrich
Digitonin, Used as non-ionic detergent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2,6-Dichloroindophenol sodium salt hydrate, suitable for vitamin C determination, BioReagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Digitonin, ~50% (TLC)
Sucrose, European Pharmacopoeia (EP) Reference Standard
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%