Merck
  • Successive determination of urinary bilirubin and creatinine employing simultaneous injection effective mixing flow analysis.
Successive determination of urinary bilirubin and creatinine employing simultaneous injection effective mixing flow analysis.
Talanta (2014-12-02)
Kraingkrai Ponhong, Norio Teshima, Kate Grudpan, Jitlada Vichapong, Shoji Motomizu, Tadao Sakai
ABSTRACT
A novel four-channel simultaneous injection effective mixing flow analysis (SIEMA) system has been assembled for successive determination of bilirubin and creatinine in urinary samples. The chemical variables and physical parameters in the flow system were optimized for the enhancement of successive analytical performances. The interferences from urine matrices on the determination of bilirubin and creatinine were eliminated to dilute urine samples. The calibration graphs with the optimum conditions were achieved to be in 0.024-5.0 mg L(-1) for bilirubin and 2-100 mg L(-1) for creatinine. The relative standard deviations (RSDs) at 3 mg L(-1) of bilirubin and at 50 mg L(-1) of creatinine for 11 runs were 1.5 and 1.0%, respectively. The limits of detections (3σ of blank) for bilirubin and creatinine were 7 µg L(-1) and 0.6 mg L(-1), respectively. The sample throughput for stepwise detection was 22 h(-1). The proposed method was applied to the successive determination of bilirubin and creatinine in urine samples.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Picric acid solution, 1.3% in H2O (saturated)
Supelco
Picric acid solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
USP
Sulfanilic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Supelco
Sulfanilic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sulfanilic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sulfanilic acid, ACS reagent, 99%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%