Merck
  • Memory B cells contribute to rapid Bcl6 expression by memory follicular helper T cells.
Memory B cells contribute to rapid Bcl6 expression by memory follicular helper T cells.
Proceedings of the National Academy of Sciences of the United States of America (2014-07-30)
Wataru Ise, Takeshi Inoue, James B McLachlan, Kohei Kometani, Masato Kubo, Takaharu Okada, Tomohiro Kurosaki
ABSTRACT
In primary humoral responses, B-cell lymphoma 6 (Bcl6) is a master regulator of follicular helper T (TFH) cell differentiation; however, its activation mechanisms and role in memory responses remain unclear. Here we demonstrate that survival of CXCR5(+) TFH memory cells, and thus subsequent recall antibody response, require Bcl6 expression. Furthermore, we show that, upon rechallenge with soluble antigen Bcl6 in memory TFH cells is rapidly induced in a dendritic cell-independent manner and that peptide:class II complexes (pMHC) on cognate memory B cells significantly contribute to this induction. Given the previous evidence that antigen-specific B cells residing in the follicles acquire antigens within minutes of injection, our results suggest that memory B cells present antigens to the cognate TFH memory cells, thereby contributing to rapid Bcl6 reexpression and differentiation of the TFH memory cells during humoral memory responses.
Sigma-Aldrich
Tamoxifen, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Tamoxifen, analytical standard