Merck
  • Regulation of CYP27B1 and CYP24A1 hydroxylases limits cell-autonomous activation of vitamin D in dendritic cells.
Regulation of CYP27B1 and CYP24A1 hydroxylases limits cell-autonomous activation of vitamin D in dendritic cells.
European journal of immunology (2014-03-20)
Rhiannon Kundu, Benjamin M Chain, Anna K Coussens, Bernard Khoo, Mahdad Noursadeghi
ABSTRACT
The active vitamin D metabolite 1α,25-dihydroxyvitamin D (1,25[OH]₂ D) potently inhibits DC priming of T-cell activation, suggesting that it mediates a homeostatic role in this context. Therefore, careful regulation of 1,25[OH]₂ D levels is necessary to avoid inappropriate inhibition of T-cell activation. Cell-autonomous control of vitamin D activity can be modulated by the action of the vitamin D-activating and -inactivating hydroxylases, CYP27B1, and CYP24A1, respectively. We show that in comparison to macrophages, human monocyte-derived DCs exhibit significantly less activation of 25-dihydroxyvitamin D to 1,25[OH]₂ D, and that DCs predominantly express a truncated CYP27B1 transcript that may contribute to the deficiency in activation of vitamin D. Furthermore, in response to stimulation with 1,25[OH]₂ D, upregulation of the inactivating enzyme CYP24A1 curtailed the functional effects of vitamin D in DCs, but not macrophages. Production of 1,25[OH]₂ D by macrophages was adequate to induce expression of vitamin D-responsive genes by DCs, inhibit DC maturation in response to innate immune stimulation and DC-dependent T-cell responses. Our data suggest that in comparison to macrophages, differential regulation of hydroxylases limits autocrine vitamin D activity in DCs, and that paracrine activation of vitamin D exerts a more potent mechanism for homeostatic control of DC function.
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material