Merck
  • G-protein receptor kinase 4 polymorphism and response to antihypertensive therapy.
G-protein receptor kinase 4 polymorphism and response to antihypertensive therapy.
Clinical chemistry (2014-10-11)
Anne M Muskalla, Paolo M Suter, Matthias Saur, Albina Nowak, Martin Hersberger, Pierre-Alexandre Krayenbuehl
ABSTRACT
G-protein receptor kinase 4 polymorphism influences blood pressure regulation via modulation of dopamine receptor D1 in renal proximal tubular cells. We investigated the role of G-protein receptor kinase 4 polymorphism in the response to hypertensive therapy in patients with essential hypertension. In a prospective study, we assessed the G-protein receptor kinase 4 polymorphisms R65L, A142V, and A486V in 100 hypertensive patients. We analyzed the association of the 3 gene variants on blood pressure control and response to antihypertensive therapy with single-locus analysis, haplotype analysis, and regression analysis. Hypertensive individuals with a homozygous double variant of 65 L and 142 V needed significantly more antihypertensive treatment (number of antihypertensives 2.59 vs 1.95, P = 0.043) and especially diuretic therapy (0.82 vs 0.49, P = 0.029) to reach the same mean arterial blood pressure than did homozygous carriers of only 1 variant or heterozygous/wild-type carriers of R65L, A142V, and A486V alleles. G-protein receptor kinase 4 polymorphism is associated with antihypertensive treatment response in patients with essential hypertension. Determination of G-protein receptor kinase 4 polymorphism may improve individual antihypertensive blood pressure control in patients with essential hypertension.
Product Description
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)