Merck
  • Solid lipid nanoparticles for potential doxorubicin delivery in glioblastoma treatment: preliminary in vitro studies.
Solid lipid nanoparticles for potential doxorubicin delivery in glioblastoma treatment: preliminary in vitro studies.
Journal of pharmaceutical sciences (2014-05-16)
Luigi Battaglia, Marina Gallarate, Elena Peira, Daniela Chirio, Elisabetta Muntoni, Elena Biasibetti, Maria Teresa Capucchio, Alberto Valazza, Pier Paolo Panciani, Michele Lanotte, Davide Schiffer, Laura Annovazzi, Valentina Caldera, Marta Mellai, Chiara Riganti
ABSTRACT
The major obstacle to glioblastoma pharmacological therapy is the overcoming of the blood-brain barrier (BBB). In literature, several strategies have been proposed to overcome the BBB: in this experimental work, solid lipid nanoparticles (SLN), prepared according to fatty acid coacervation technique, are proposed as the vehicle for doxorubicin (Dox), to enhance its permeation through an artificial model of BBB. The in vitro cytotoxicity of Dox-loaded SLN has been measured on three different commercial and patient-derived glioma cell lines. Dox was entrapped within SLN thanks to hydrophobic ion pairing with negatively charged surfactants, used as counterions. Results indicate that Dox entrapped in SLN maintains its cytotoxic activity toward glioma cell lines; moreover, its permeation through hCMEC/D3 cell monolayer, assumed as a model of the BBB, was increased when the drug was entrapped in SLN. In conclusion, SLN proved to be a promising vehicle for the delivery of Dox to the brain in glioblastoma treatment.
Sigma-Aldrich
Sodium 1-decanesulfonate, ≥99.0% (T)
Sigma-Aldrich
Sodium palmitate, ≥98.5%
Sigma-Aldrich
Sodium 1-decanesulfonate, ~98%
Supelco
Sodium 1-decanesulfonate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sodium stearate, ≥99%
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sucrose, European Pharmacopoeia (EP) Reference Standard
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%