Merck
  • Cholecystokinin octapeptide regulates the differentiation and effector cytokine production of CD4(+) T cells in vitro.
Cholecystokinin octapeptide regulates the differentiation and effector cytokine production of CD4(+) T cells in vitro.
International immunopharmacology (2014-04-08)
Jing-Ge Zhang, Jun-Xu Liu, Xian-Xian Jia, Jing Geng, Feng Yu, Bin Cong
ABSTRACT
Cholecystokinin octapeptide (CCK-8), an immunomodulatory peptide, can promote or suppress the development or function of specific CD4(+) T cell subsets by regulating antigen-presenting cell functions. In the current study, we investigated whether CCK-8 exerts a direct effect on T cells through influencing differentiation and cytokine production of distinct CD4(+) T cell subsets in vitro. Our results showed that CCK-8 differentially affects the development and function of CD4(+) T cell populations, with a negative influence on Th1 and Th17 cells and positive regulatory effect on inducible T regulatory cells (iTreg). Notably, CCK-8 suppressed Th1 while slightly enhancing Th2 development and cytokine production. Similarly, CCK-8 inhibited the differentiation of Th17 cells and promoted Foxp3 expression. L-364,718 and LY-288,513, selective antagonists of CCK1R and CCK2R, respectively, suppressed the effects of CCK-8 on CD4(+) T cell subset-specific transcription factors. Our findings strongly indicate that CCK-8 exerts a direct effect on T cells, which is dependent on CCKRs, particularly CCK2R. The collective results aid in further clarifying the mechanism underlying the anti-inflammatory and immunoregulatory effects of CCK-8.
Sigma-Aldrich
Rat IL-4 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IL-4 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Cotton Rat IL4 / Interleukin 4 ELISA Kit
Sigma-Aldrich
Mouse IL-4 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human IL-4 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Mouse IL-4 ELISA Kit, for serum, plasma and cell culture supernatant