Merck
The chemistry of graphene oxide.
Chemical Society reviews (2009-12-22)
Daniel R Dreyer, Sungjin Park, Christopher W Bielawski, Rodney S Ruoff
ABSTRACT
The chemistry of graphene oxide is discussed in this critical review. Particular emphasis is directed toward the synthesis of graphene oxide, as well as its structure. Graphene oxide as a substrate for a variety of chemical transformations, including its reduction to graphene-like materials, is also discussed. This review will be of value to synthetic chemists interested in this emerging field of materials science, as well as those investigating applications of graphene who would find a more thorough treatment of the chemistry of graphene oxide useful in understanding the scope and limitations of current approaches which utilize this material (91 references).
Sigma-Aldrich
Potassium chlorate, puriss., 99-101%
Sigma-Aldrich
Potassium chlorate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium chlorate, ACS reagent, ≥99.0%