Merck
  • Peroxisomal Atg37 binds Atg30 or palmitoyl-CoA to regulate phagophore formation during pexophagy.
Peroxisomal Atg37 binds Atg30 or palmitoyl-CoA to regulate phagophore formation during pexophagy.
The Journal of cell biology (2014-02-19)
Taras Y Nazarko, Katharine Ozeki, Andreas Till, Geetha Ramakrishnan, Pouya Lotfi, Mingda Yan, Suresh Subramani
ABSTRACT
Autophagy is a membrane trafficking pathway that sequesters proteins and organelles into autophagosomes. The selectivity of this pathway is determined by autophagy receptors, such as the Pichia pastoris autophagy-related protein 30 (Atg30), which controls the selective autophagy of peroxisomes (pexophagy) through the assembly of a receptor protein complex (RPC). However, how the pexophagic RPC is regulated for efficient formation of the phagophore, an isolation membrane that sequesters the peroxisome from the cytosol, is unknown. Here we describe a new, conserved acyl-CoA-binding protein, Atg37, that is an integral peroxisomal membrane protein required specifically for pexophagy at the stage of phagophore formation. Atg30 recruits Atg37 to the pexophagic RPC, where Atg37 regulates the recruitment of the scaffold protein, Atg11. Palmitoyl-CoA competes with Atg30 for Atg37 binding. The human orthologue of Atg37, acyl-CoA-binding domain containing protein 5 (ACBD5), is also peroxisomal and is required specifically for pexophagy. We suggest that Atg37/ACBD5 is a new component and positive regulator of the pexophagic RPC.
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥0.5 unit/mg solid
Sigma-Aldrich
Rabbit IgG−Agarose, saline suspension
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Avanti
16:0 Coenzyme A, Avanti Research - A Croda Brand 870716P, powder
Sigma-Aldrich
Anti-ACBD5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Millipore
EZview Red Anti-HA Affinity Gel
Sigma-Aldrich
Human IgG−Agarose, suspension
Sigma-Aldrich
Leupeptin trifluoroacetate salt, ≥90% (HPLC), microbial
Roche
X-tremeGENE 9 DNA Transfection Reagent, Polymer reagent for transfecting common cell lines