Merck
  • Inhibition of the TRAIL death receptor by CMV reveals its importance in NK cell-mediated antiviral defense.
Inhibition of the TRAIL death receptor by CMV reveals its importance in NK cell-mediated antiviral defense.
PLoS pathogens (2014-08-15)
Shilpi Verma, Andrea Loewendorf, Qiao Wang, Bryan McDonald, Alec Redwood, Chris A Benedict
ABSTRACT
TNF-related apoptosis inducing ligand (TRAIL) death receptors (DR) regulate apoptosis and inflammation, but their role in antiviral defense is poorly understood. Cytomegaloviruses (CMV) encode many immune-modulatory genes that shape host immunity, and they utilize multiple strategies to target the TNF-family cytokines. Here we show that the m166 open reading frame (orf) of mouse CMV (MCMV) is strictly required to inhibit expression of TRAIL-DR in infected cells. An MCMV mutant lacking m166 expression (m166stop) is severely compromised for replication in vivo, most notably in the liver, and depleting natural killer (NK) cells, or infecting TRAIL-DR-/- mice, restored MCMV-m166stop replication completely. These results highlight the critical importance for CMV to have evolved a strategy to inhibit TRAIL-DR signaling to thwart NK-mediated defenses.
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)