Merck
  • TMEFF2 deregulation contributes to gastric carcinogenesis and indicates poor survival outcome.
TMEFF2 deregulation contributes to gastric carcinogenesis and indicates poor survival outcome.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-07-06)
Tiantian Sun, Wan Du, Hua Xiong, Yanan Yu, Yurong Weng, Linlin Ren, Huijun Zhao, Yingchao Wang, Yingxuan Chen, Jie Xu, Yongbing Xiang, Wenxin Qin, Weibiao Cao, Weiping Zou, Haoyan Chen, Jie Hong, Jing-Yuan Fang
ABSTRACT
The role and clinical implication of the transmembrane protein with EGF and two follistatin motifs 2 (TMEFF2) in gastric cancer is poorly understood. Gene expression profile analyses were performed and Gene Set Enrichment Analysis (GSEA) was used to explore its gene signatures. AGS and MKN45 cells were transfected with TMEFF2 or control plasmids and analyzed for gene expression patterns, proliferation, and apoptosis. TMEFF2 expression was knocked down with shRNAs, and the effects on genome stability were assessed. Interactions between TMEFF2 and SHP-1 were determined by mass spectrometry and immunoprecipitation assays. Integrated analysis revealed that TMEFF2 expression was significantly decreased in gastric cancer cases and its expression was negatively correlated with the poor pathologic stage, large tumor size, and poor prognosis. GSEA in The Cancer Genome Atlas (TCGA) and Jilin datasets revealed that cell proliferation, apoptosis, and DNA damage-related genes were enriched in TMEFF2 lower expression patients. Gain of TMEFF2 function decreased cell proliferation by increasing of apoptosis and blocking of cell cycle in gastric cancer cells. The protein tyrosine phosphatase SHP-1 was identified as a binding partner of TMEEF2 and mediator of TMEFF2 function. TMEFF2 expression positively correlated with SHP-1, and a favorable prognosis was more likely in patients with gastric cancer with higher levels of both TMEFF2 and SHP-1. TMEFF2 acts as a tumor suppressor in gastric cancer through direct interaction with SHP-1 and can be a potential biomarker of carcinogenesis.
Sigma-Aldrich
MISSION® esiRNA, targeting human PTPN6
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sirpa
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human SIRPA
Sigma-Aldrich
MISSION® esiRNA, targeting human NR0B2
Sigma-Aldrich
MISSION® esiRNA, targeting human TMEFF2
Sigma-Aldrich
Anti-TMEFF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ptpn6
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ala-D-γ-Glu-Lys-D-Ala-D-Ala, ≥97% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining