Merck
  • The comparative effects of perindopril and catechin on mesangial matrix and podocytes in the streptozotocin induced diabetic rats.
The comparative effects of perindopril and catechin on mesangial matrix and podocytes in the streptozotocin induced diabetic rats.
Pharmacological reports : PR (2014-06-10)
Salime Pelin Ertürküner, Murat Başar, Matem Tunçdemir, İsmail Seçkin
ABSTRACT
Hyperglycemia and advanced glucose end substance (AGE) are responsible for excessive reactive oxygen species (ROS) production, which causes oxidative stress in diabetes mellitus. Oxidative stress and high blood pressure may cause injury and glomerulosclerosis in the kidney. End-stage kidney failure induced by glomerulosclerosis leads to microalbuminuria (Ma) in diabetic nephropathy. We investigated the effects of an angiotensin converting enzyme inhibitor (ACEI), perindopril, and an antioxidant, catechin, on podocytes and the glomerular mesangial matrix in experimental diabetic nephropathy using ultrastructural visualization and immunohistochemical staining. We compared 5 groups of male adult Wistar albino rats: a control group, an untreated diabetic group, and diabetic groups treated with perindopril, catechin, or catechin+perindopril. Blood glucose values in all diabetic groups were significantly higher than in the control group (p < 0.001). The body weight in all diabetic groups was significantly lower than in the control group (p < 0.001, p < 0.05). The kidney weight in the catechin+perindopril-treated diabetic group was significantly lower than in the untreated diabetic group (p < 0.001). In all treated diabetic groups, Ma levels decreased significantly (p < 0.001). Mesangial matrix and podocyte damage increased in the untreated diabetic group, but the group treated with catechin+perindopril showed less damage. TGF-beta 1 immunostaining was significantly lower in the catechin-treated and perindopril-treated groups than in the untreated diabetic group (p < 0.001). Catechin was more effective than ACEI in preventing podocyte structure. Podocytes appeared to be the first cells affected in diabetes mellitus. When exposed to hyperglycemia, podocytes caused the mesangial matrix to expand. Catechin and perindopril were more effective in preventing renal corpuscle damage when administered together.
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Ethanol, anhydrous, denatured
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Grids for transmission electron microscopy, grid size 300 mesh × 83 μm pitch, copper
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%